1-Methyl-1H-indazol-6-boronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Methyl-1H-indazole-6-boronic Acid, 1-Methylindazole-6-boronic acid
430480010
ACRO430480010EA
212
EUR
ACRO430480010
1-Methyl-1H-indazol-6-boronsäure 97%
1-Methyl-1H-indazole-6-boronic acid
Formel:
C₈H₉BN₂O₂ Molecular Weight: 175,9803 g/mol Schmelzpunkt: 259,8…261.2 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09870053 CAS-Nummer: 1150114-80-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Enantiomeric excess | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...