2-Methyl-2H-indazol-6-boronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2-Methyl-2H-indazol-6-yl)boronic acid
430490010
ACRO430490010EA
263
EUR
ACRO430490010
2-Methyl-2H-indazol-6-boronsäure 97%
2-Methyl-2H-indazole-6-boronic acid
Formel:
C₈H₉BN₂O₂ Molecular Weight: 175,9800 g/mol Schmelzpunkt: 306…311 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09870057 CAS-Nummer: 1001907-57-8 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Specific optical rotation | 3.0° to 5.0° (20°C, 589 nm) (c=1, chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...