1H-Indazol-4-boronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(1H-Indazol-4-yl)boronic acid, 1H-indazole-4-boronic acid
430500010
ACRO430500010EA
171
EUR
ACRO430500010
1H-Indazol-4-boronsäure 97%
1H-Indazole-4-boronic acid
Formel:
C₇H₇BN₂O₂ Molecular Weight: 161,9500 g/mol Schmelzpunkt: 166…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878901 CAS-Nummer: 1023595-17-6 |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 165 to 169 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...