1H-Benzimidazol-5-boronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

430540010
ACRO430540010EA 151 EUR
ACRO430540010
1H-Benzimidazol-5-boronsäurepinakolester 97%
1H-Benzimidazol-5-boronsäurepinakolester
Formel: C₁₃H₁₇BN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11054041
CAS-Nummer: 1007206-54-3

Bestellen

Spezifikation Testergebnisse

Appearance White to tan to purple Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR