Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,1,3-Benzothiadiazole-5-boronic acid, pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole, Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester

430570010
ACRO430570010EA 171 EUR
ACRO430570010
Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester 97%
Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester
Formel: C₁₂H₁₅BN₂O₂S
Molecular Weight: 262,134 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11867874
CAS-Nummer: 1168135-03-2

Bestellen

Spezifikation Testergebnisse

Appearance Brown Powder
Infrared spectrum Conforms
Melting point 83 to 87 °C
GC ≥96.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR