3-(Brommethyl)phenylboronsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxy(3-(brommethyl)phenyl)boran, (3-(Brommethyl)phenyl)metaborsäure, 3-(Brommethyl)benzolboronsäure

430600010 430600050
ACRO430600010EA 37 EUR
ACRO430600010 ACRO430600050
3-(Brommethyl)phenylboronsäure 97%
3-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
Molecular Weight: 214.85
Schmelzpunkt: 209…211 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01632207
CAS-Nummer: 51323-43-4

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.5 %
Color scale ≤20 APHA
Acidity ≤0.001 meq/g
Alkalinity ≤0.002 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Halogenated compounds ≤0.001 % (as Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR