5-Fluor-2-methylphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

430650050
ACRO430650050EA 164 EUR
ACRO430650050
5-Fluor-2-methylphenylboronsäure 97%
5-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03095047
CAS-Nummer: 163517-62-2

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 154 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR