2-Formylbenzeneboronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

430670010 430670050
ACRO430670010EA 41.3 EUR
ACRO430670010 ACRO430670050
2-Formylbenzeneboronic acid pinacol ester 97%
2-Formylbenzeneboronic acid pinacol ester
Formel: C₁₃H₁₇BO₃
Lagertemperatur: Kühlschrank
MDL: MFCD07363841
CAS-Nummer: 380151-85-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to dark yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR