1H-Indazol-6-boronsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

431020010
ACRO431020010EA 228 EUR
ACRO431020010
1H-Indazol-6-boronsäure 97%
1H-Indazole-6-boronic acid
Formel: C₇H₇BN₂O₂
Schmelzpunkt: 180…184 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04972419
CAS-Nummer: 885068-10-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR