8-Isochinolincarbonsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Isoquinoline-8-carboxylic acid

431050010
ACRO431050010EA 229 EUR
ACRO431050010
8-Isochinolincarbonsäure 97%
8-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
Molecular Weight: 173,1681 g/mol
Schmelzpunkt: 263…267 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06738935
CAS-Nummer: 61563-43-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 263 to 267 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR