4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carboxaldehyde, 4H,5H,6H,7H-PYRAZOLO[1,5-A]PYRIDINE-3-CARBALDEHYDE

431090010
ACRO431090010EA 162 EUR
ACRO431090010
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde 97%
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde
Formel: C₈H₁₀N₂O
Molecular Weight: 150,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834817
CAS-Nummer: 307308-03-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to orange Low melting solid or liquid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR