Ethyl-6-chloro-3-pyridazinecarboxylate 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 6-chloro-3-pyridazinecarboxylate, Ethyl 6-chloropyridazine-3-carboxylate, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, 6-Chloro-pyridazine-3-carboxylic acid ethyl ester
431110010
431110050
ACRO431110010EA
160
EUR
ACRO431110010
ACRO431110050
Ethyl-6-chloro-3-pyridazinecarboxylate 95%
Ethyl-6-chloro-3-pyridazinecarboxylate
Formel:
C₇H₇ClN₂O₂ Molecular Weight: 186.595 g/mol Schmelzpunkt: 149…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09908168 CAS-Nummer: 75680-92-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 148 to 153 °C |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...