4-Methylcyclohexen-1-ylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Methylcyclohex-1-en-1-yl)boronic acid, 4-Methyl-1-cyclohexen-1-ylboronic acid
431430010
ACRO431430010EA
213
EUR
ACRO431430010
4-Methylcyclohexen-1-ylboronsäure 97%
4-Methylcyclohexene-1-ylboronic acid
Formel:
C₇H₁₃BO₂ Molecular Weight: 139,99 g/mol Schmelzpunkt: 155…159 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06659857 CAS-Nummer: 850567-92-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...