Natrium-3-(trimethylsilyl)propionat-[D4] (98% D)

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, Sodium 3-(trimethylsilyl)propionate-[²H₄], 3-(Trimethylsilyl)propionic acid-d4 sodium salt

432120010
ACRO432120010EA 163 EUR
ACRO432120010
Natrium-3-(trimethylsilyl)propionat-[D4] (98% D)
Natrium-3-(trimethylsilyl)propionat-[D4]
Formel: C₆H₉D₄NaO₂Si
Molecular Weight: 172,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002762
CAS-Nummer: 24493-21-8

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
D-Enrichment ≥97.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR