Chlorodimethyl-3,3,3-trifluoropropylsilane 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(3,3,3-Trifluoropropyl)chlorodimethylsilane
432210050
ACRO432210050EA
130
EUR
ACRO432210050
Chlorodimethyl-3,3,3-trifluoropropylsilane 95%
Chlorodimethyl-3,3,3-trifluoropropylsilane
Formel:
C₅H₁₀ClF₃Si Molecular Weight: 190,67 g/mol Dichte: 1,11 g/cm³ Flammpunkt: 20 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053211 CAS-Nummer: 1481-41-0 UN: 2986 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to straw yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3720 to 1.3780 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...