N'-Hydroxypyridine-4-carboximidamide 97%

Lieferant: Thermo Fisher Scientific

432480050
ACRO432480050EA 192 EUR
ACRO432480050
N'-Hydroxypyridine-4-carboximidamide 97%

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline solid
Infrared spectrum Conforms
Melting point 200 to 207 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR