2-Amino-4-phenylthiazol 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
432490010
432490050
ACRO432490010EA
45.9
EUR
ACRO432490010
ACRO432490050
2-Amino-4-phenylthiazol 97%
2-Amino-4-phenylthiazole
Formel:
C₉H₈N₂S Schmelzpunkt: 150…152,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039680 CAS-Nummer: 2010-06-2 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 149 to 153 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...