3-Bromo-5-(trifluoromethyl)pyridin-2-ol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-bromo-5-(trifluoromethyl)pyridin-2-ol, 3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine, 3-bromo-5-(trifluoromethyl)pyridine-2-ol

432560050
ACRO432560050EA 302 EUR
ACRO432560050
3-Bromo-5-(trifluoromethyl)pyridin-2-ol 97%
3-Bromo-5-(trifluoromethyl)pyridin-2-ol
Formel: C₆H₃BrF₃NO
Molecular Weight: 241,995 g/mol
Schmelzpunkt: 170,1…171.2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02691223
CAS-Nummer: 76041-73-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR