5-Fluor-2-hydroxypyridin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
432580050
432580250
ACRO432580050EA
118
EUR
ACRO432580050
ACRO432580250
5-Fluor-2-hydroxypyridin 97%
5-Fluor-2-hydroxypyridin
Formel:
C₅H₄FNO Schmelzpunkt: 152…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092918 CAS-Nummer: 51173-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...