trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
trans-4-(Trifluoromethyl)cyclohexane-carboxylicacid
432600050
ACRO432600050EA
568
EUR
ACRO432600050
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid
Formel:
C₈H₁₁F₃O₂ Molecular Weight: 196,17 g/mol Schmelzpunkt: 153…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037363 CAS-Nummer: 133261-33-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 152 to 157 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...