2-(Trifluormethyl)pyrazin 97%
Lieferant: Thermo Scientific
|
Achtung
|
432610050
ACRO432610050EA
290
EUR
ACRO432610050
2-(Trifluormethyl)pyrazin 97%
2-(Trifluoromethyl)pyrazine
Formel:
C₅H₃F₃N₂ Molecular Weight: 148,09 g/mol Siedepunkt: 116…118 °C (760 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797734 CAS-Nummer: 61655-67-2 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...