6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Bromo[1,3]oxazolo[4,5-b]pyridin-2(3H)-one, 6-bromooxazolo[4,5-b]pyridin-2(3H)-one

432680050
ACRO432680050EA 494 EUR
ACRO432680050
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one 97%
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one
Formel: C₆H₃BrN₂O₂
Molecular Weight: 215,006 g/mol
Schmelzpunkt: 230…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689590
CAS-Nummer: 21594-52-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Melting point 229 to 236 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR