Imidazo[1,2-a]pyridin-2-carbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

432780010 432780050
ACRO432780010EA 80.3 EUR
ACRO432780010 ACRO432780050
Imidazo[1,2-a]pyridin-2-carbonsäure 97%
Imidazo[1,2-a]pyridin-2-carbonsäure
Formel: C₈H₆N₂O₂·xH₂O
Molecular Weight: 162.15 g/mol (anhy)
Schmelzpunkt: 232…234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17171427
CAS-Nummer: 64951-08-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Additional info Hydrate, contains 9.5% to 11.0% water
HPLC ≥96.0 %
Elemental Analysis 14.96 to 16.17 % N
Residue after evaporation ≤0.01 %
Peroxides ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR