Diphenyliodoniumtrifluormethansulfonat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
432840010
ACRO432840010EA
96.9
EUR
ACRO432840010
Diphenyliodoniumtrifluormethansulfonat 99%
Diphenyliodonium trifluoromethanesulphonate
Formel:
C₁₃H₁₀F₃IO₃S Schmelzpunkt: 179…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191356 CAS-Nummer: 66003-76-7 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 176 to 180 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...