4-Aminopiperidin 95%

Lieferant: Thermo Fisher Scientific
Gefahr

432920050 432920250
ACRO432920050EA 88 EUR
ACRO432920050 ACRO432920250
4-Aminopiperidin 95%
4-Aminopiperidin
Formel: C₅H₁₂N₂
Siedepunkt: 65 °C (18Torr)
Dichte: 0,954 g/cm³
Flammpunkt: 46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179399
CAS-Nummer: 13035-19-3
UN: 2733
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR