Ethyl-2-hydrazinylacetate hydrochloride 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl hydrazinoacetate hydrochloride, Ethyl 2-hydrazinylacetate Hydrochloride, Ethyl hydrazinoacetate HCl, Ethyl 2-hydrazinylacetate hydrochloride

433150100
ACRO433150100EA 265 EUR
ACRO433150100
Ethyl-2-hydrazinylacetate hydrochloride 97%
Ethyl-2-hydrazinylacetate hydrochloride
Formel: C₄H₁₁ClN₂O₂
Molecular Weight: 154,594 g/mol
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012923
CAS-Nummer: 6945-92-2

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to light orange Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 146 to 151 °C
Titration Argentometric ≥96.0 to ≤104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR