(3,5-Dichlorphenyl)methansulfonsäurechlorid 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,5-Dichloro-α-toluenesulphonyl chloride
433230010
ACRO433230010EA
222
EUR
ACRO433230010
(3,5-Dichlorphenyl)methansulfonsäurechlorid 97%
(3,5-Dichlorophenyl)methanesulphonyl chloride
Formel:
C₇H₅CI₃O₂S Molecular Weight: 259,54 g/mol Schmelzpunkt: 108,6…110.6 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631928 CAS-Nummer: 163295-70-3 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 106 to 112 °C |
HPLC | ≥96.0 % as diethylsulfonamide derivative |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...