Dextransulfat Natriumsalz, M.W. ∼ 500,000

Lieferant: Thermo Fisher Scientific

433240050 433240250 433241000
ACRO433240050EA 59.2 EUR
ACRO433240050 ACRO433240250 ACRO433241000
Dextransulfat Natriumsalz, M.W. ∼ 500,000
Dextran sulphate sodium salt
Formel: (C₆H₇Na₃O₁₄S₃)n
Lagertemperatur: Kühlschrank
MDL: MFCD00081551
CAS-Nummer: 9011-18-1

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light yellow Powder
Loss on drying ≤10 %
Sulfated ash 40 % to 50 %
Solubility (10 % in water) Clear to slightly hazy colorless to light yellow
Molecular weight ≥500000
pH 5 to 7.5 (1 % in water)
Specific optical rotation +75° to +105° (20°C, 589 nm) (c=6, water)
Specific viscosity 0.10 to 0.70 (1.0M in NaCl, 25°C)
Sulfur (S) 17 to 20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR