7-Amino-1-tetralon 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Amino-alpha-tetralone, 7-Amino-3,4-dihydro-1(2H)-naphthalenone

433260010 433260050
ACRO433260010EA 83.5 EUR
ACRO433260010 ACRO433260050
7-Amino-1-tetralon 97%
7-Amino-1-tetralon
Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07369817
CAS-Nummer: 22009-40-1

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 135 to 140 °C
GC ≥96.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR