Guanidiniumthiocyanat 99+% für die Molekularbiologie

Lieferant: Thermo Fisher Scientific
Gefahr

433941000
ACRO433941000EA 82 EUR
ACRO433941000
Guanidiniumthiocyanat 99+% für die Molekularbiologie
Guanidiniumthiocyanat
Formel: NH₂C(=NH)NH₂·HSCN
Schmelzpunkt: 118…122 °C
Dichte: 1,29 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013027
CAS-Nummer: 593-84-0
UN: 3261
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 118 to 122 °C
Titration Argentometric 99.0 to 101.0 %
Loss on drying ≤0.5 %
Sulfated ash ≤0.05 %
Solubility (3 M in water) Clear colorless
UV (3 M in water)
pH 5.0 to 7.0 (1 M in water)
Dnase,Rnase,Protease activity None detected
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Zinc (Zn) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR