3,6-Dibromcarbazol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,6-Dibromo-9H-carbazole
433970050
ACRO433970050EA
77.5
EUR
ACRO433970050
3,6-Dibromcarbazol 97%
3,6-Dibromocarbazole
Formel:
C₁₂H₇Br₂N Molecular Weight: 325 g/mol Schmelzpunkt: 212…214 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004961 CAS-Nummer: 6825-20-3 |
Spezifikation Testergebnisse
Appearance | Tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 211 to 215 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...