(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 97%
Lieferant: Thermo Scientific
Synonyme:
(2S,5S)-(-)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 2-{2-[(2S,5S)-2,5-Diethylphospholan-1-yl]phenyl}-1,3-dioxolane, 2-{2-[(2S,5S)-2,5-Diethyl-1-phospholano]phenyl}1,3-dioxolane
434121000
ACRO434121000EA
42
EUR
ACRO434121000
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 97%
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane
Formel:
C₁₇H₂₅O₂P Molecular Weight: 292,36 g/mol Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09265099 CAS-Nummer: 1314246-02-0 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
NMR | ≥96.0 % |
GC | ≥99.5 % |
Residue after evaporation | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...