5-Brom-2-iodpyrimidin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
434250010
ACRO434250010EA
156
EUR
ACRO434250010
5-Brom-2-iodpyrimidin 97%
5-Bromo-2-iodopyrimidine
Formel:
C₄H₂BrIN₂ Schmelzpunkt: 100…102 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01318111 CAS-Nummer: 183438-24-6 |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 99 to 103 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...