Ammoniumsulfat 99%, reinst

Lieferant: Thermo Fisher Scientific

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

434380010 434380050
ACRO434380010EA 38.9 EUR
ACRO434380010 ACRO434380050
Ammoniumsulfat 99%, reinst
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
Merck Index: 13,00559

Bestellen

Spezifikation Testergebnisse

Appearance White Crystals
Sulfated ash ≤0.05 %
pH 5.0 to 6.0
Titration excess NaOH ≥98.5 %
Chloride (Cl) ≤0.003 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR