(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Tarenflurbil, (R)-2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, (R)-(-)-Flurbiprofen

434490010
ACRO434490010EA 180 EUR
ACRO434490010
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure 97%
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244.260 g/mol
Schmelzpunkt: 106…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00869714
CAS-Nummer: 51543-40-9
UN: 2811
ADR: 6.1 II

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 106 to 110 °C
HPLC ≥96.0 %
Specific optical rotation -35° to -45° (20 °C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR