Tris-acetat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate

434500250 434501000 434505000
ACRO434500250EA 56.1 EUR
ACRO434500250 ACRO434501000 ACRO434505000
Tris-acetat 99%
Tris-acetat
Formel: NH₂C(CH₂OH)₃·CH₃COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD00038951
CAS-Nummer: 6850-28-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Water ≤1.0 %
Solubility (1 g/ml in water) Clear colorless
UV at 290 nm A: ≤0.5
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR