2-(2,2,2-Trifluoracetyl)thiophen 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Trifluoroacetyl)thiophene

434700050
ACRO434700050EA 105 EUR
ACRO434700050
2-(2,2,2-Trifluoracetyl)thiophen 98%
2-(2,2,2-Trifluoracetyl)thiophen
Formel: C₆H₅ClOS
Molecular Weight: 180.15 g/mol
Flammpunkt: 59 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00041427
CAS-Nummer: 651-70-7
UN: 1993
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR