tert-Butyl-N-allylcarbamate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate
434770100
434770500
ACRO434770100EA
60.2
EUR
ACRO434770100
ACRO434770500
tert-Butyl-N-allylcarbamate 98%
tert-Butyl-N-allylcarbamate
Formel:
C₈H₁₅NO₂ Siedepunkt: 48…50 °C (1 mmHg) Schmelzpunkt: 34,9…38.3 °C Flammpunkt: 79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191870 CAS-Nummer: 78888-18-3 |
Spezifikation Testergebnisse
Appearance | Colorless to light yellow Low melting solid |
Infrared spectrum | Conforms |
Melting point | 33 to 40 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...