4-Brom-m-toluylsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Brom-3-methylbenzoesäure
434780050
434780250
ACRO434780050EA
61.2
EUR
ACRO434780050
ACRO434780250
4-Brom-m-toluylsäure 97%
4-Brom-m-toluylsäure
Formel:
C₈H₇BrO₂ Schmelzpunkt: 215,5…216.4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039526 CAS-Nummer: 7697-28-1 |
Spezifikation Testergebnisse
Appearance | Yellow to orange to tan Powder |
Infrared spectrum | Conforms |
Melting point | 213 to 217 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...