Carbobenzyloxy-beta-Alanin 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Carbobenzyloxy-beta-alanine

434810050 434810250
ACRO434810050EA 50.9 EUR
ACRO434810050 ACRO434810250
Carbobenzyloxy-beta-Alanin 98%
Carbobenzyloxy-beta-Alanin
Formel: C₁₁H₁₃NO₄
Molecular Weight: 223.23 g/mol
Schmelzpunkt: 105.3…107.2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037292
CAS-Nummer: 2304-94-1

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Melting point 100 to 110 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR