3-Cyanophenylacetic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

434890010 434890050
ACRO434890010EA 124 EUR
ACRO434890010 ACRO434890050
3-Cyanophenylacetic acid 95%
3-Cyanophenylacetic acid
Formel: C₉H₇NO₂
Schmelzpunkt: 117…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06411079
CAS-Nummer: 1878-71-3
UN: 3439
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 113 to 119 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR