(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane 97%

Lieferant: Thermo Scientific

Synonyme: (2S,5S)-(+)-1-(2-(1,3-Dioxolan-2-yl)phenyl-2,5-dimethylphospholane, 2-{2-[(2S,5S)-2,5-Dimethylphospholan-1-yl]phenyl}-1,3-dioxolane, (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane

435171000
ACRO435171000EA 47.6 EUR
ACRO435171000
(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane 97%
(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Formel: C₁₅H₂₁O₂P
Molecular Weight: 264,305 g/mol
Dichte: 1,103 g/cm³
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09265097
CAS-Nummer: 695816-47-8

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
NMR ≥96.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR