1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: [(R,R)Ethyl-DuPhosRh(COD)]BF4, (-)-1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, (-)-1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(l) tetrafluoroborate, (R,R)-Et-DUPHOS-Rh, 1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

435182500
ACRO435182500EA 236 EUR
ACRO435182500
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Formel: C₃₀H₄₈BF₄P₂Rh
Molecular Weight: 660,371 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862464
CAS-Nummer: 228121-39-9

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red to brown Powder
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure
NMR (19F-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 52.4 to 56.8 % C
Specific optical rotation +40° to +50° (20°C, 589 nm) (c=1 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR