1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(1Z,5Z)-cycloocta-1,5-diene, (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethyl-phospholane, rhodium, tetrafluoroborate, (S,S)-ME-BPE-RH, (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, ME-BPE-RH
435235000
ACRO435235000EA
404
EUR
ACRO435235000
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
Formel:
C₂₂H₃₈BF₄P₂Rh Molecular Weight: 554,19 g/mol Schmelzpunkt: 152…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038435 CAS-Nummer: 213343-65-8 |
Spezifikation Testergebnisse
Appearance | Orange to red to brown Crystalline powder |
Infrared spectrum | Conforms |
NMR (1H-NMR) | Consistent with structure |
NMR (19F-NMR) | Consistent with structure |
NMR (31P-NMR) | Consistent with structure |
Specific optical rotation | +35° to +45° (20°C, 589 nm) (c=1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...