4-Fluor-3-methylphenylboronsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid

435330010 435330050 435330250
ACRO435330010EA 16 EUR
ACRO435330010 ACRO435330050 ACRO435330250
4-Fluor-3-methylphenylboronsäure 98%
4-Fluor-3-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
Schmelzpunkt: 215.4…217.3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863527
CAS-Nummer: 139911-27-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR