4-Fluor-3-methylphenylboronsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid
435330010
435330050
435330250
ACRO435330010EA
16
EUR
ACRO435330010
ACRO435330050
ACRO435330250
4-Fluor-3-methylphenylboronsäure 98%
4-Fluor-3-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153.95 g/mol Schmelzpunkt: 215.4…217.3 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863527 CAS-Nummer: 139911-27-6 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...