6-Indolcarbaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Indol-6-carboxaldehyd

435510010
ACRO435510010EA 220 EUR
ACRO435510010
6-Indolcarbaldehyd 97%
6-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179596
CAS-Nummer: 1196-70-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Crystalline powder
Infrared spectrum Conforms
Melting point 127 to 131 °C
HPLC ≥96.0 %
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR