3-Aminophenylboronsäure Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

435530010 435530050
ACRO435530010EA 68.3 EUR
ACRO435530010 ACRO435530050
3-Aminophenylboronsäure Hydrochlorid 98%
3-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00191748
CAS-Nummer: 85006-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR