(R)-tert-Butyl(1,4-dihydroxybutan-2-yl)carbamate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-2-Boc-Amino-Butane-1,4-Diol, (R)-tert-Butyl (1,4-dihydroxybutan-2-yl)carbamate, (R)-(+)-2-(BOC-Amino)-1,4-butanediol
435570050
ACRO435570050EA
163
EUR
ACRO435570050
(R)-tert-Butyl(1,4-dihydroxybutan-2-yl)carbamate 97%
(R)-tert-Butyl(1,4-dihydroxybutan-2-yl)carbamate
Formel:
C₉H₁₉NO₄ Molecular Weight: 205,25 g/mol Schmelzpunkt: 66,4…72.1 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08703679 CAS-Nummer: 397246-14-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 65 to 69 °C |
GC | ≥96.0 % |
Enantiomeric excess | ≥98.0 % |
Specific optical rotation | +7° to +9° (20 °C, 589 nm) (c=1, chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...