Chlor(diethyl)isopropylsilan 95+%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Diethylisopropylsilyl Chloride
435650250
ACRO435650250EA
717
EUR
ACRO435650250
Chlor(diethyl)isopropylsilan 95+%
Chlorodiethylisopropylsilane
Formel:
C₇H₁₇ClSi Molecular Weight: 164,75 g/mol Dichte: 0,9 g/cm³ Flammpunkt: 38 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00191631 CAS-Nummer: 107149-56-4 UN: 2986 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4370 to 1.4390 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...